Learn More
Saint Gobain Performance Plastics™ Versilic Silicone Tubing
Brand:  Saint Gobain Performance Plastics™ 760440
Product Code. 10151201
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Density 1.2 translucent temperature range -60°C to 200°C, reel of 25m, 5mm bore X 10mm O.D. Versilic™