missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Versican Antibody (S351-23), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22408JF549
This item is not returnable.
View return policy
Description
Versican Monoclonal antibody specifically detects Versican in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Versican | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Chondroitin sulfate proteoglycan 2WGN, Chondroitin sulfate proteoglycan core protein 2, CSPG2DKFZp686K06110, ERVR, GHAP, Glial hyaluronate-binding protein, Large fibroblast proteoglycan, PG-MWGN1, versican, versican core protein, versican proteoglycan | |
| Fusion protein amino acids 362-585 (glycosaminoglycan alpha domain) of mouse Versican core protein | |
| 0.1 mL | |
| Extracellular Matrix | |
| 1462 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| S351-23 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction