Learn More
Thermo Scientific™ VersaTREK™ REDOX™ EZ Draw™ Media
Perform safe, simple and convenient phlebotomy, without the need for costly blood adapters, using the only FDA-cleared blood culture bottles for true direct draws.
Specifications
| Content And Storage | Store away from direct sunlight |
|---|---|
| Test Type | Microbial Detection |
| No. per Pack | 50 Bottles/Box |
| Quantity | 40 mL |
| Specimen Type | Blood, Sterile Body Fluid |
Description
Perform safe, simple and convenient phlebotomy, without the need for costly blood adapters, with Thermo Scientific™ VersaTREK™ REDOX™ EZ Draw™ Media, the only FDA-cleared true, direct measured draw in the industry. Accommodating up to 5mL of blood, EZ Draw media is available in a 40mL broth for both REDOX 1 aerobic and REDOX 2 anaerobic bottles and can be used for manual or automated monitoring of direct-draw blood samples. Plus, the highly-enriched REDOX media supports a wide range of organisms, enhancing the fully automated VersaTREK System's ability to better recover fastidious organisms.
Meet the demands of both high and low volume laboratories with the Thermo Scientific VersaTREK System, which comes with a full line of ancillary and accessory products. With four FDA-cleared tests and a simple, two-bottle media system, the VersaTREK System provides maximum value with faster results and fewer limitations.- Easy and convenient sampling
- Reduced risk from sharps by eliminating the need for syringe, butterfly, angel wings and other blood transfer devices
- Reduced costs by eliminating the need for separate collection devices
- Largest blood-to-broth dilution ratio in the industry (1:9) to dilute out the effects of antimicrobials, serum host factors and other agents
- Enhanced, enriched media to support the wide range of organisms found in the microbiology laboratory
- Available in 40 and 80mL sizes and suitable for all patient populations
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
| Store away from direct sunlight | |
| 50 Bottles/Box | |
| Blood, Sterile Body Fluid |
| Microbial Detection | |
| 40 mL | |
| 0.1 to 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.