missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ VersaTouch™ 23-201 Series Blue PVC Gloves

Product Code. p-8005385 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
6 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15361005 10 6 pairs
15341005 8 6 pairs
15331005 7 6 pairs
15351005 9 6 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15361005 Supplier Ansell™ Supplier No. 23201100
Add to basket
Add to basket
This item is not returnable. View return policy

Comfort long sleeve PVC glove with good dexterity and pliability even at low temperature.

  • Rough sandblast finish provide good wet-grip
  • Soft PVC coating on glove maintains dexterity and pliabiity at low temperatures
  • Dipped rough finish makes handling wet and slippery materials easier and safer
  • Heat sealed sleeve offers liquid resistance and dust protection only

Applications:

  • Chemical handling
  • Chemical refining
  • Cleaning
  • Construction work
  • Dairy processing
  • Fish processing
  • Fruit and vegetable processing
  • General purpose
  • Janitorial use
  • Light assembly tasks
  • Maintenance
  • Refining oil and petrol
TRUSTED_SUSTAINABILITY

Specifications

Back Material PVC
Certifications/Compliance Standards CATEGORY III, CE Mark, EN 374, EN 388 : 4121
Cuff Material PVC
Applications Industrial
Coating Material PVC
Color Blue
Latex Free Yes
Material PVC
Product Type Chemical-resistant Gloves
Size 10
Thickness (Metric) 4 mm
Glove Cuff Style Heat-sealed Sleeve
Lining Cotton
Quantity 72 Pairs
Resistant To Chemical and liquid
Series 23-201
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.