missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Versapor™ Acrylic Copolymer 5000 Membrane Disc Filter

Product Code. 17174491
Change view
Click to view available options
Diameter (Metric):
37 mm
47 mm
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
17174491 47 mm Pack of 100
17194551 37 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17174491 Supplier Cytiva (formerly Pall Lab) Supplier No. 60178
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Disc Filters useful for prefiltration of particle-laden samples, serum filtration, and dissolution testing.

  • Protects instrumentation against particulate build up
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY

Specifications

Material Versapor
Pore Size 5 μm
Sterility Non-sterile
Thickness (Metric) 94 μm
Diameter (Metric) 47 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.