Learn More
3M™ Versaflo™ TR-600 PAPR Accessory, Filter
3M Versaflo™ TR-600 PAPR Accessory, Filter is used with 3M™ Versaflo™ TR-600 series air purifying device.
Brand:  3M™ 7100020971
29.75 GBP valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.31500kg
 View more versions of this product
Product Code. 15779067
  • £40.25 / Each
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
Description
Description
  • Simple to install
  • For use with 3M Versaflo Powered Air TR-600 series
  • Recommends use of TR-6100FC filter cover to help protect filter from physical damage
Specifications
Specifications
Particulate Filter
3M Versaflo Powered Air Turbo TR-600 Series
TR-6710E