Learn More
3M™ Versaflo™ Powered Air Turbo Starter Kit, TR-619UK
Click to view available options
:
Dimensions (L x W x H): 0.346 x 0.346 x 0.254 m
Description
- Powered air turbo kit that keeps workers comfortable and protected for their work day
- Multiple airflow rate options for user comfort
- Ergonomically designed to fit close to the body and allow for greater movement in tight work spaces
- Multiple belt size adjustments for proper fit and comfort
- Belt designed with flexible air channels to minimise heat buildup
Specifications
Specifications
| Battery Type | Lithium-ion |
| Product Type | PAPR Assembly, No Headtop |
| Model | Versaflo™ |
| Dimensions (L x W x H) | 0.346 x 0.346 x 0.254 m |