Learn More
3M™ Versaflo™ Powered Air Respirator Starter Kit
Product Code. 15698566
Click to view available options
:
Model: TR315UK
Product Code. 15698566
Brand: 3M™ 7100098909
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Combines performance, protection and functionality with modern, stylish, lightweight design and unrivaled comfort. PAPR Starter Kit is a lightweight, versatile system for environments with hazardous dust and mist.
The Versaflo™ powered air system combines performance, protection and functionality with modern, stylish, lightweight design and unrivaled comfort. Lightweight, versatile respiratory protective system for environments with hazardous dust and mist.
Simple starter kit with all the component parts to get you up and running. Simply add the headtop required. Includes:
  • TR-315UK+ Turbo Unit
  • Particle Filter
  • Pre-filter
  • Standard belt
  • High-capacity battery
  • Battery charger
  • Length adjusting breathing tube
  • Air flow indicator
TRUSTED_SUSTAINABILITY
Specifications
Product Type PAPR Assembly, No Headtop
For Use With (Equipment) UK and Ireland electrical requirements
Includes TR-315UK+ Turbo unit, P filter, pre-filter, standard belt, high capacity battery, battery charger kit, and self adjusting breathing tube and air flow indicator
Model TR-315UK
Quantity 1 Ea.