missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ Peel-Off Visor Cover M-928

Product Code. 17257630
Change view
Click to view available options
:
Transparent
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17257630 Transparent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17257630 Supplier 3M™ Supplier No. 70071564341

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Self-adhesive visor cover for protecting 3M™ Versaflo™ M-Series Headtop visors from scratches and overspray of paint or chemicals.

  • Transparent, peel-off visor cover for 3M™ Versaflo™ M-Series Headtops
  • Fits over visor, protecting it from scratching or overspray of paint and chemicals
  • Self-adhesive – can be easily applied and peeled off
  • Up to five covers can be attached, one on top of the other
TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
For Use With (Application) Protecting 3 M™ Versaflo™ M-Series Headtop Visors
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.