missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ M-400 Series Respiratory Helmet Assembly

Product Code. 11793916
Change view
Click to view available options
Model:
M-407
Includes:
Flame-Resistant Shroud
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Includes unitSize
11793916 M-407 Flame-Resistant Shroud Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11793916 Supplier 3M™ Supplier No. 7000104038

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used with an approved air source, M-400 Series Helmets provide respiratory (OSHA APF=1000) and limited head (ANSI Z89.1-2003 Type 1 Class G) and eye (ANSI Z87.1-2010) protection.

Features:

  • Chemical and Scratch Resistant Lenses
  • Large Viewing Area
  • Lightweight, Easy Visor Replacement System

Environmental Conditions:

  • Cold Working Environment
  • High Mobility Required
  • Safety Helmet Required

Hazard Type:

  • Gas, Vapour and Particulates
  • Liquid Splash
  • Low Energy Flying Particles/Dust
  • Falling Objects
  • Sparks and Hot Particles

Includes:

  • The M-405 assembly includes a M-925 Visor, M-444 Inner Collar, and M-445 Outer Shroud.
    • The M-925 visor is made of polycarbonate.
    • The M-445 shroud is appropriate for general purpose use.
  • The M-407 Assembly includes a M-927 Premium Visor, M-444 Inner Collar, and M-447 Flame Resistant Outer Shroud.
    • The M-927 visor is made of coated polycarbonate.
    • The M-447 shroud is made of Nomex flame resistant fibers.
  • 3M Catalog# M-405 , 3M Catalog# M-407
TRUSTED_SUSTAINABILITY

Specifications

Assigned Protection Factor (APF) 1,000
Certifications/Compliance CE Approved, EN 12941 TH3, EN 14594 3B, EN 166:2001, EN 397
Color Black
Material Polyaramid
Product Type Respirator Helmet
For Use With (Application) Demolition, Metalworking, Metal Processing, Smelting, Foundries (Grinding, Polishing), Woodworking, Construction
For Use With (Equipment) 3M Air Delivery Units
Includes Flame-Resistant Shroud
Material (Lens) Polycarbonate
Model M-407
Sterility Non-sterile
Suspension Type Textile
Visor Type Polycarbonate
With Faceseal No
Dimensions (L x W x H) 346 x 346 x 254 mm
Length (English) 13.6 in.
Length (Metric) 346 mm
Width (English) 13.6 in.
Width (Metric) 346 mm
Height (English) 9.6 in.
Height (Metric) 254 mm
Weight (English) 2.4 lb.
Weight (Metric) 1066 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.