Learn More
3M™ Versaflo™ Inner Collar, M-444
Product Code. 17207660
Change view
Click to view available options
:
3M™ Versaflo™ M-400 Headtop
Product Code.
17207660 3M™ Versaflo™ M-400 Headtop
1 options
Product Code. 17207660 Supplier 3M™ Supplier No. 7100137832
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Replacement Inner Collar for use with 3M™ Versaflo™ M-400 Headtop. The Inner Collar zips into the shroud and provides a comfortable respiratory seal.
  • Replacement inner collar
  • Provides a comfortable respiratory seal
  • For use 3M™Versaflo™ M-400 Series Helmets
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene Coated Knitted Polyamid
For Use With (Equipment) 3 M™ Versaflo™ M-400 Headtop