Learn More
3M™ Versaflo™ Industrial Helmet with Hood Powered Air Respirator Kit, TR-300E+ IHK
Product Code. 31297044 Shop All 3M Products
Change view
Click to view available options
Size:
One Size
Product Code. Size
31297044 One Size
1 options
Product Code. 31297044 Supplier 3M™ Supplier No. 7100318453
  • ONLINE EXCLUSIVE PRICE
    £1745.00
    £1088.17 / Pack of 1
    Save £656.83
    37% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Versatile and easy-to-use respiratory protection against particulates hazards.
  • TR-300+ Powered Air Respirator Systems are engineered for versatility and customisation and offer a wide range of headtop, breathing tubes, and accessories to create a customised Powered Air Respirator solution.
TRUSTED_SUSTAINABILITY
Specifications
Color White
Product Type PAPR Assembly with Hood
Size One Size