Learn More
3M™ Versaflo™ Cleaning and Storage Replacement Seals, TR-654
Click to view available options
:
Model: TR-654
Description
- Seals and plugs protect powered air turbo TR-600 Series from internal contamination during cleaning
- Enable powered air turbo unit TR-600 Series to be rinsed or submerged
- Protect inlet and outlet openings of the unit during storage
- All plug seals should be replaced after 30 uses or if they appear worn or damaged
Specifications
Specifications
| Certifications/Compliance | CE Approved |
| Product Type | TR-654 Seals |