missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Verekitug Humanized Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555024
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA.
Verekitug inhibits the activity of specific enzymes involved in the replication of viral RNA, thereby preventing the virus from multiplying.
Specifications
| Verekitug Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ILXR | |
| Recombinant Human TSLPR/CRLF2. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 1 mg/mL | |
| PBS with no preservative | |
| Human | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction