missing translation for 'onlineSavingsMsg'
Learn More

Verdiperstat, MedChemExpress

Product Code. 16497438
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497438 10 mg 10mg
16479917 5 mg 5mg
16443467 50 mg 50mg
16437416 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497438 Supplier Medchem Express Supplier No. HY17646
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Verdiperstat (AZD3241) is a selective, irreversible and orally active myeloperoxidase (MPO) inhibitor, with an IC50 of 630 nM, and can be used in the research of neurodegenerative brain disorders.

Verdiperstat (AZD3241) is a selective, irreversible and orally active myeloperoxidase (MPO) inhibitor, with an IC50 of 630 nM, and can be used in the research of neurodegenerative brain disorders.

Specifications

Chemical Name or Material Verdiperstat
CAS 890655-80-8
Color White
Purity Grade Notes Research
Molecular Formula C11H15N3O2S
Quantity 10 mg
Synonym AZD3241
Solubility Information DMSO : 150 mg/mL (592.14 mM; Need ultrasonic)
SMILES O=C(N1)C(NC=C2)=C2N(CCOC(C)C)C1=S
Molecular Weight (g/mol) 253.32
Formula Weight 253.32
Percent Purity 99.6%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.