missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Verbascoside

Product Code. 16494727
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494727 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16494727 Supplier MedChemExpress Supplier No. HYN0021
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Verbascoside is isolated from Lantana camara, acts as an ATP-competitive inhibitor of PKC, with an IC50 of 25 μM, and has antitumor, anti-inflammatory and antineuropathic pain activity.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 624.59 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 61276-17-3
Product Type Natural Products
Purity 95.67%
Solubility DMSO : ≥ 100 mg/mL (160.11 mM)
Synonym Acteoside∣ Kusaginin∣ TJC160
Target PKC∣Apoptosis∣Bacterial∣HSV
Quantity 50 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.