missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Veratryl alcohol, MedChemExpress
Description
Veratryl alcohol (3,4-Dimethoxybenzenemethanol), a secondary metabolite of some lignin degrading fungi, is commonly used nonphenolic substrate for assaying ligninolytic activity.
Specifications
Specifications
| Chemical Name or Material | Veratryl alcohol |
| CAS | 93-03-8 |
| Color | Colorless to Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C9H12O3 |
| Quantity | 100 mg |
| Synonym | 3,4-Dimethoxybenzyl alcohol |
| Solubility Information | Ethanol : 100 mg/mL (594.57 mM; Need ultrasonic) |
| SMILES | OCC1=CC=C(OC)C(OC)=C1 |
| Molecular Weight (g/mol) | 168.19 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?