missing translation for 'onlineSavingsMsg'
Learn More

Veratrole, 99%, Thermo Scientific Chemicals

Catalog Number 11454024
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
91-16-7
C8H10O2
138.166
MFCD00008357
ABDKAPXRBAPSQN-UHFFFAOYSA-N
veratrole, veratrol, pyrocatechol dimethyl ether, o-dimethoxybenzene, catechol dimethyl ether, benzene, 1,2-dimethoxy, 2-methoxyanisole, o,o-dimethyl catechol, 2-dimethoxybenzol, benzene, o-dimethoxy
7043
CHEBI:59114
1,2-dimethoxybenzene
COC1=CC=CC=C1OC
This item is not returnable. View return policy
91-16-7
C8H10O2
138.166
MFCD00008357
ABDKAPXRBAPSQN-UHFFFAOYSA-N
veratrole, veratrol, pyrocatechol dimethyl ether, o-dimethoxybenzene, catechol dimethyl ether, benzene, 1,2-dimethoxy, 2-methoxyanisole, o,o-dimethyl catechol, 2-dimethoxybenzol, benzene, o-dimethoxy
7043
CHEBI:59114
1,2-dimethoxybenzene
COC1=CC=CC=C1OC

It is useful for organic synthesis of aromatic compounds. Veratrole is relatively electron-rich and thus readily undergoes electrophilic substitution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is useful for organic synthesis of aromatic compounds. Veratrole is relatively electron-rich and thus readily undergoes electrophilic substitution.

Solubility
Soluble in alcohol, diethyl ether, acetone, and methanol. Slightly soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 21°C to 23°C
Density 1.084
Boiling Point 206°C to 207°C
Flash Point 87°C (188°F)
Refractive Index 1.533
Quantity 250 g
Beilstein 1364621
Merck Index 14,9956
Solubility Information Soluble in alcohol,diethyl ether,acetone,and methanol. Slightly soluble in water.
Formula Weight 138.17
Percent Purity 99%
Chemical Name or Material Veratrole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'