missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Veratridine
GREENER_CHOICE

Product Code. 18702674
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18702674

Brand: Tocris Bioscience™ 2918/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Voltage-gated Na+ channel opener

Voltage-gated Na+ channel opener; increases intracellular Ca2+ with no effect on the Na+/Ca2+ exchanger. Steroid-derived alkaloid neurotoxin. Increases membrane Na+ permeability and depolarizes excitable tissues.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-62-5
Molecular Formula C36H51NO11

Specifications

Quantity 50 mg
Formula Weight 673.79
Purity >98%
Molecular Weight (g/mol) 673.8
Chemical Name or Material Veratridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.