missing translation for 'onlineSavingsMsg'
Learn More

Veratraldehyde, 99%

Product Code. 11418993
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11418993

Brand: Thermo Scientific Alfa Aesar A11110.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-14-9 | C9H10O3 | 166.18 g/mol

Veratraldehyde was used in the preparation of 4-chloromethyl-2-(dimethoxyphenyl)-1,3-dioxolane. It was used in the synthesis of (+)-lithospermic acid, having anti-HIV activity. It forms 1:1 inclusion complexes with cyclodextrins. It reacts with 3-acetyl-2,5-dimethythiophene to yield chalcone dye, (2E)-3-(3,4-Dimethoxyphenyl)-1-(2,5-dimethylthiophen-3-yl)prop-2-en-1-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Veratraldehyde was used in the preparation of 4-chloromethyl-2-(dimethoxyphenyl)-1,3-dioxolane. It was used in the synthesis of (+)-lithospermic acid, having anti-HIV activity. It forms 1:1 inclusion complexes with cyclodextrins. It reacts with 3-acetyl-2,5-dimethythiophene to yield chalcone dye, (2E)-3-(3,4-Dimethoxyphenyl)-1-(2,5-dimethylthiophen-3-yl)prop-2-en-1-one.

Solubility
Soluble in water. Freely soluble in alcohol and ether.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-14-9
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00003363
InChI Key WJUFSDZVCOTFON-UHFFFAOYSA-N
Synonym veratraldehyde, veratric aldehyde, methylvanillin, vanillin methyl ether, veratral, veratryl aldehyde, benzaldehyde, 3,4-dimethoxy, veratrum aldehyde, p-veratric aldehyde, 4-o-methylvanillin
PubChem CID 8419
ChEBI CHEBI:17098
IUPAC Name 3,4-dimethoxybenzaldehyde
SMILES COC1=CC=C(C=O)C=C1OC

Specifications

Melting Point 42°C to 46°C
Boiling Point 281°C
Flash Point 112°C (233°F)
Quantity 100 g
Beilstein 473899
Sensitivity Air Sensitive
Merck Index 14,9951
Solubility Information Soluble in water. Freely soluble in alcohol and ether.
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material Veratraldehyde
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.