missing translation for 'onlineSavingsMsg'
Learn More

(±)-Verapamil Hydrochloride, MP Biomedicals™

Product Code. 11425502 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425502 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11425502 Supplier MP Biomedicals Supplier No. 0219554501
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Functions as a calcium channel modulator, adrenoceptor antagonist, anti-arrhythmic, cardiac depressant, and coronary vasodilator. (±)-Verapamil Hydrochloride, MP Biomedicals is an α1-antagonist and calcium channel-blocker that acts by inhibit

Acts by plugging up the channels and limiting the entry of calcium into both smooth muscle cells of arteriolar walls and the cardiac muscle cells at higher doses.

  • Blocks L-type Ca2+ channels in smooth and cardiac muscle
  • Induces apoptosis of human primary and metastatic colon adenocarcinoma cells in vitro
  • Drug resistance reversal agent acting on Pgp, e.g. decrease renal tubule elimination of digoxin
  • Increases basal ATPase activity of Pgp
  • Substrate of Cyp3A4 and CYP2C6

Specifications

CAS 152-11-4
Melting Point 140°C to 150°C (literature)
Molecular Formula C27H39ClN2O4
Quantity 1 g
Synonym verapamil hydrochloride, verapamil hcl, manidon, calcan hydrochloride, cardibeltin, cardiabeltin, cardioprotect, veroptinstada, calaptin, caveril
Solubility Information Soluble in water (7g/100mL); sparingly soluble in chloroform; soluble in ethanol (26mg/mL), isopropanol, acetone, ethyl acetate; freely soluble in methanol (>100mg/mL), DMF (>100mg/mL); practically insoluble in ether.
InChI Key DOQPXTMNIUCOSY-UHFFFAOYSA-N
SMILES CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC.Cl
IUPAC Name 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride
Molecular Weight (g/mol) 491.069
PubChem CID 62969
Formula Weight 491.1
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.