missing translation for 'onlineSavingsMsg'
Learn More

Verapamil hydrochloride, MedChemExpress

Product Code. 16440718
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16440718 5 g 5g
16412009 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16440718 Supplier Medchem Express Supplier No. HYA0064
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Verapamil hydrochloride ((±)-Verapamil hydrochloride) is a calcium channel blocker and a potent and orally active first-generation P-glycoprotein (P-gp) inhibitor. Verapamil hydrochloride also inhibits CYP3A4. Verapamil hydrochloride has the potential for high blood pressure, heart arrhythmias and angina research.

Verapamil hydrochloride ((±)-Verapamil hydrochloride) is a calcium channel blocker and a potent and orally active first-generation P-glycoprotein (P-gp) inhibitor. Verapamil hydrochloride also inhibits CYP3A4. Verapamil hydrochloride has the potential for high blood pressure, heart arrhythmias and angina research.

Specifications

Chemical Name or Material Verapamil hydrochloride
CAS 152-11-4
Color White
Purity Grade Notes Research
Molecular Formula C27H39ClN2O4
Quantity 5 g
Synonym (±)-Verapamil hydrochlorideCP-16533-1 hydrochloride
Solubility Information H2O : 50 mg/mL (101.82 mM; Need ultrasonic) ∣DMSO : ≥ 31 mg/mL (63.13 mM)
SMILES [H]Cl.COC1=CC=C(C(C#N)(C(C)C)CCCN(CCC2=CC=C(OC)C(OC)=C2)C)C=C1OC
Molecular Weight (g/mol) 491.06
Formula Weight 491.06
Percent Purity 99.01%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.