missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Verapamil hydrochloride, 99+%

Product Code. 15464119
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15464119 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15464119 Supplier Thermo Scientific Alfa Aesar Supplier No. J61535.03
  • ONLINE EXCLUSIVE PRICE
    £61.51
    £51.65 / 1g
    Save £9.86
    16% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 152-11-4 | C27H39ClN2O4 | 491.069 g/mol

±)-Verapamil hydrochloride is a calcium channel modulator, adrenoceptor antagonist, anti-arrhythmic, cardiac depressant, and coronary vasodilator. It is also a calcium channel-blocker. It acts by inhibiting the slow channel entry of calcium into the cell. It acts by plugging up the channels and limiting the entry of calcium into both smooth muscle cells of arteriolar walls and the cardiac muscle cells at higher doses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
±)-Verapamil hydrochloride is a calcium channel modulator, adrenoceptor antagonist, anti-arrhythmic, cardiac depressant, and coronary vasodilator. It is also a calcium channel-blocker. It acts by inhibiting the slow channel e
TRUSTED_SUSTAINABILITY

Specifications

CAS 152-11-4
Melting Point 138°C to 141°C
Color White
Molecular Formula C27H39ClN2O4
MDL Number MFCD00055208
Quantity 1 g
UN Number UN2811
Beilstein 3647093
Merck Index 14,9950
Synonym verapamil hydrochloride, verapamil hcl, manidon, calcan hydrochloride, cardibeltin, cardiabeltin, cardioprotect, veroptinstada, calaptin, caveril
Solubility Information Soluble in water; Also soluble in ethanol or methanol
InChI Key DOQPXTMNIUCOSY-UHFFFAOYSA-N
SMILES CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC.Cl
IUPAC Name 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride
Molecular Weight (g/mol) 491.069
PubChem CID 62969
Formula Weight 491.07
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material (+/-)-Verapamil hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.