missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Verapamil EP Impurity C (as Hydrochloride), Mikromol™
Description
Verapamil EP Impurity C (as Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | 2-(3,4-Dimethoxyphenyl)-N,N-dimethylethan-amine Hydrochloride |
| CAS | 51012-67-0 |
| Analyte or Component Names | 2-(3,4-Dimethoxyphenyl)-N,N-dimethylethan-amine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H19 N O2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-(3,4-Dimethoxyphenyl)-N,N-dimethylethanamine Hydrochloride, Verapamil Hydrochloride Imp. C (EP), Verapamil Imp. C (EP), 2-(3,4-Dimethoxyphenyl)-N,N-dimethylethan-amine Hydrochloride, Verapamil Hydrochloride Imp. C (EP) as Hydrochloride, Verapamil Hydrochloride Impurity C as Hydrochloride, Verapamil Impurity C as Hydrochloride |
| InChI Formula | InChI=1S/C12H19NO2.ClH/c1-13(2)8-7-10-5-6-11(14-3)12(9-10)15-4;/h5-6,9H,7-8H2,1-4H3;1H |
| SMILES | CN(C)CCC1=CC=C(OC)C(OC)=C1.[H]Cl |
| IUPAC Name | 2-(3,4-dimethoxyphenyl)-N,N-dimethylethanamine;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?