missing translation for 'onlineSavingsMsg'
Learn More

Veralipride, MedChemExpress

Product Code. 16486297
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486297 5 mg 5mg
16469986 50 mg 50mg
16454686 100 mg 100mg
16443578 10 mg 10mg
16400066 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16486297 Supplier Medchem Express Supplier No. HY101797
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Veralipride is a D2 receptor antagonist. It is an alternative antidopaminergic treatment for menopausal symptoms.

Veralipride is a D2 receptor antagonist. It is an alternative antidopaminergic treatment for menopausal symptoms.

Specifications

Chemical Name or Material Veralipride
CAS 66644-81-3
Color White
Purity Grade Notes Research
Molecular Formula C17H25N3O5S
Quantity 5 mg
Synonym (±)-Veralipride LIR166
Solubility Information DMSO : ≥ 100 mg/mL (260.78 mM)
SMILES O=C(NCC1N(CC=C)CCC1)C2=CC(S(=O)(N)=O)=CC(OC)=C2OC
Molecular Weight (g/mol) 383.46
Formula Weight 383.46
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.