Learn More
Emma Safety Footwear Vera XD Safety Shoes, Yellow
Product Code. 15908611
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15908611 37 1 pair
15918611 38 1 pair
15928611 39 1 pair
15938611 40 1 pair
15948611 41 1 pair
15958611 42 1 pair
15968611 43 1 pair
15978611 44 1 pair
15988611 45 1 pair
15998611 46 1 pair
15908621 47 1 pair
15918621 48 1 pair
15928621 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15908611 Supplier Emma Safety Footwear Supplier No. MM50753437
Only null left
Add to basket
Add to basket
An easy shoe for wipe clean hygiene. The upper is fully washable unique microfiber and features Hydro-Tec™ Sanitized Silver for dry, fresh feet. The protective toe cap prevents staining and is lubricant and fat resistant. Developed for use on hard tiled floors and other easy-to-clean floors.
  • S2-Safety shoe for wet working conditions
  • Microfiber construction
  • GripForce™ Duo Heat Resistant Outsole
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Design Vera XD, Yellow
Material Microfiber
Size 37