Learn More
Emma Safety Footwear Vera Safety Shoes, Green
An easy shoe for wipe clean hygiene. The upper is fully washable unique microfiber and features Hydro-Tec™ Sanitized Silver for dry, fresh feet. The protective toe cap prevents staining and is lubricant and fat resistant. Developed for use on hard tiled floors and other easy-to-clean floors.
Brand: Emma Safety Footwear MM50650438
Additional Details : Weight : 1.26300kg
Description
- S2-Safety shoe for wet working conditions
- GripForce™ Duo Outsole
- Microfiber construction
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver
Specifications
EN ISO 20345:2011 | |
White | |
Hydro-Tec™ Sanitized Silver | |
38 |
Elastic | |
Vera, Green | |
Microfiber | |
Steel |