missing translation for 'onlineSavingsMsg'
Learn More

VER-50589, MedChemExpress

Product Code. 16461589
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461589 1 mg 1mg
16449497 5 mg 5mg
16406958 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16461589 Supplier Medchem Express Supplier No. HY15984
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress VER-50589 is a Hsp90 inhibitor, with an IC50 of 21 nM and a Kd of 4.5 nM.

VER-50589 is a Hsp90 inhibitor, with an IC50 of 21 nM and a Kd of 4.5 nM.

Specifications

Chemical Name or Material VER-50589
CAS 747413-08-7
Color White
Purity Grade Notes Research
Molecular Formula C19H17ClN2O5
Quantity 1 mg
Solubility Information DMSO : ≥ 48 mg/mL (123.46 mM)
SMILES O=C(C1=NOC(C2=CC(Cl)=C(O)C=C2O)=C1C3=CC=C(OC)C=C3)NCC
Molecular Weight (g/mol) 388.8
Formula Weight 388.8
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.