missing translation for 'onlineSavingsMsg'
Learn More

VER-49009, MedChemExpress

Product Code. 16490266
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490266 2 mg 2mg
16479497 5 mg 5mg
16436958 10 mg 10mg
16417036 100 mg 100mg
16403017 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16490266 Supplier Medchem Express Supplier No. HY15986
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress VER-49009 is a Hsp90 inhibitor, with an IC50 of 25 nM and a Kd of 78 nM.

VER-49009 is a Hsp90 inhibitor, with an IC50 of 25 nM and a Kd of 78 nM.

Specifications

Chemical Name or Material VER-49009
CAS 558640-51-0
Color White
Purity Grade Notes Research
Molecular Formula C19H18ClN3O4
Quantity 2 mg
Synonym CCT 129397
Solubility Information DMSO : 100 mg/mL (257.85 mM; Need ultrasonic)
SMILES O=C(C1=NNC(C2=CC(Cl)=C(O)C=C2O)=C1C3=CC=C(OC)C=C3)NCC
Molecular Weight (g/mol) 387.82
Formula Weight 387.82
Percent Purity 99.39%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.