missing translation for 'onlineSavingsMsg'
Learn More

VER-155008, MedChemExpress

Product Code. 16485216
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485216

Brand: Medchem Express HY10941

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VER-155008 is an inhibitor of Hsp70, with IC50s of 0.5 μM, 2.6 μM, and 2.6 μM for Hsp70, Hsc70 and Grp7, respectively, and with a Kd of 0.3 μM for Hsp70.

VER-155008 is an inhibitor of Hsp70, with IC50s of 0.5 μM, 2.6 μM, and 2.6 μM for Hsp70, Hsc70 and Grp7, respectively, and with a Kd of 0.3 μM for Hsp70.

Specifications

Chemical Name or Material VER-155008
CAS 1134156-31-2
Color Pink
Purity Grade Notes Research
Molecular Formula C25H23Cl2N7O4
Quantity 100 mg
Solubility Information DMSO : ≥ 37 mg/mL (66.50 mM)
SMILES O[C@@H]([C@H]([C@H](N1C(NCC2=CC=C(Cl)C(Cl)=C2)=NC3=C1N=CN=C3N)O4)O)[C@H]4COCC5=CC=C(C#N)C=C5
Molecular Weight (g/mol) 556.4
Formula Weight 556.4
Percent Purity 97.25%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.