missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEPH1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94141-0.02ml
This item is not returnable.
View return policy
Description
VEPH1 Polyclonal antibody specifically detects VEPH1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| VEPH1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ12604, KIAA1692MGC111426, MELT, MGC126709, MGC142115, Protein melted, ventricular zone expressed PH domain homolog 1 (zebrafish), ventricular zone-expressed PH domain-containing protein homolog 1, VEPH | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 448-547 of human VEPH1 (NP_078897.2). KSLAFHTMLTKGVGSDDGEDENRGDIPASISLSEIDPLGQGNDKLPFKTDTERSQLGESSVSYPNIIHIDSENLSETVKENSQEETPETTASPIEYQDKL | |
| 0.02 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79674 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction