Learn More
Veolia Water Systems Veolia Water Systems™ Sterilization Tablet
Product Code. 10542572
Change view
Click to view available options
Quantity:
12/Pk.
Product Code. Quantity
10542572 12/Pk.
1 options
Product Code. 10542572 Supplier Veolia Water Systems Supplier No. CT1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Reliable and effective chlorine tablets that reduce bio film and protect the system against bacterial growth.
  • CT1 Tablets (pack of 12) - Chlorine tablets
  • Sanitizes tubing or pipe work to remove bacteria
  • Reduces bio film and protect the system against bacterial growth
  • Sterilization tablet
Specifications
Quantity 12/Pk.