Learn More
Veolia Water Systems™ Deionization Cartridge
Product Code. 11924867
Click to view available options
For Use With (Equipment):
MAXIMA MK1/MK2
MEDICA 7/15 Mk1, Option 1/2/3/4/5/6, Purelab Option 7/15 and PS
Unit Size:
Each
Product Code. 11924867
Brand: Veolia Water Systems™ LC126
Only null left
Add to basket
Add to basket
Robust and reliable water purification cartridges ideal for removing ionic contaminants to achieve specified water quality.
  • Deionization cartridges
  • Removes ionic contaminants to achieve specified water quality
  • Compatible with Maxima MK1/MK2
Specifications
For Use With (Equipment) MAXIMA MK1/MK2