missing translation for 'onlineSavingsMsg'
Learn More

VENTX Antibody, DyLight 755, Novus Biologicals™

Product Code. 18427672 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427672 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427672 Supplier Novus Biologicals Supplier No. NBP177192IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VENTX Polyclonal antibody specifically detects VENTX in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen VENTX
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias homeobox protein VENTX, HPX42BNA88A, MGC119910, MGC119911, VENT homeobox homolog, VENT homeobox homolog (Xenopus laevis), VENT-like homeobox 2, VENT-like homeobox protein 2, VENTX2hemopoietic progenitor homeobox protein VENTX2
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the center of human VENTX. The immunogen is located within amino acids 120 - 170 of VENTX. Amino Acid Squence: CFQNRRMKHKRQMQDPQ
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27287
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.