missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VENTX Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77192-0.025mg
This item is not returnable.
View return policy
Description
VENTX Polyclonal specifically detects VENTX in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| VENTX | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| homeobox protein VENTX, HPX42BNA88A, MGC119910, MGC119911, VENT homeobox homolog, VENT homeobox homolog (Xenopus laevis), VENT-like homeobox 2, VENT-like homeobox protein 2, VENTX2hemopoietic progenitor homeobox protein VENTX2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 27287 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 μg/mL | |
| NP_055283 | |
| VENTX | |
| Antibody was raised against a 16 amino acid synthetic peptide near the center of human VENTX. The immunogen is located within amino acids 120 - 170 of VENTX. Amino Acid Squence: CFQNRRMKHKRQMQDPQ | |
| 0.025 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction