missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Venlafaxine hydrochloride

Product Code. 17291172 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17291172 5 g 5g
17281172 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17291172 Supplier Thermo Scientific Chemicals Supplier No. 465810050
  • ONLINE EXCLUSIVE PRICE
    £110.00
    £99.65 / 5g
    Save £10.35
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 99300-78-4 | C17H28ClNO2, C17H27NO2·HCl | 313.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99300-78-4
Molecular Formula C17H27NO2·HCl

Specifications

Quantity 5 g
Solubility Solubility aq. soln.: slightly soluble.
Melting Point 217.0°C
Formula Weight 313.86
Chemical Name or Material Venlafaxine hydrochloride
compliance-icons
Product Identifier
  • Venlafaxine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Venlafaxine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.