missing translation for 'onlineSavingsMsg'
Learn More

Venlafaxine.HCl, Lipomed

Product Code. 30459398
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30459398

Brand: Lipomed LPMVLF1063HC10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Venlafaxine.HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Venlafaxine Hydrochloride
CAS 99300-78-4
Analyte or Component Names Venlafaxine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C17 H27 N O2 . Cl H
Synonym Cyclohexanol, 1-[2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]-, hydrochloride (1:1), Cyclohexanol, 1-[2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]-, hydrochloride (9 CI), Efexor, Effexor, VENIZ-XR, VENTAB, Venlafaxin hydrochloride, Venlafaxine hydrochloride, Venlift, Ventab XL, Vexor, Wy 45030, Wy 4530
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.COc1ccc(cc1)C(CN(C)C)C2(O)CCCCC2
IUPAC Name 1-[2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol;hydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 313.86
Quantity 10 mg
Formula Weight 313.1809 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.