missing translation for 'onlineSavingsMsg'
Learn More

Venlafaxine Cyclic Impurity, TRC

Product Code. 30455705
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455705 10 mg 10mg
30451025 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30455705 Supplier Toronto Research Chemicals Supplier No. TRCV12001510MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93413-70-8
Molecular Formula C17 H25 N O2
Molecular Weight (g/mol) 275.39
Synonym Venlafaxine EP Impurity E
IUPAC Name 5-(4-methoxyphenyl)-3-methyl-1-oxa-3-azaspiro[5.5]undecane
SMILES COc1ccc(cc1)C2CN(C)COC23CCCCC3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H25NO2/c1-18-12-16(14-6-8-15(19-2)9-7-14)17(20-13-18)10-4-3-5-11-17/h6-9,16H,3-5,10-13H2,1-2H3
Formula Weight 275.1885
Chemical Name or Material Venlafaxine Cyclic Impurity H
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.