missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Venema Rolls, Qualitative and Technical Papers, Smooth, Grade 100/N

Product Code. 15902720 Shop All Sartorius Products
Click to view available options
Dimensions (L x W):
1000m x 150 mm
1000m x 240 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15902720

Brand: Sartorius Stedim Biotech™ FT13281501000

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Sartorius Stedim Biotech™ Venema Rolls, Qualitative and Technical Papers, Smooth, Grade 100/N are suitable for analytical and technical purposes and commonly used in the sugar industry.

  • Suitable for use in the sugar industry
  • Suitable for use with Venema systems
  • Wet strengthened and smooth
  • Cellulose construction
TRUSTED_SUSTAINABILITY

Specifications

Basis Weight 85 g/m2
Description Venema roll
For Use With (Application) Assaying sugar
Grade 100/N
Quantity 1 Roll
Thickness (Metric) 0.18 mm
Dimensions (L x W) 1000m x 150 mm
Length (Metric) 1000 m
Width (Metric) 150 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.