missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vemurafenib, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
Description
Vemurafenib (PLX4032) is a first-in-class, selective, potent inhibitor of B-RAF kinase, with IC50s of 31 and 48 nM for RAFV600E and c-RAF-1, respectively. Vemurafenib induces cell autophagy.
Specifications
Specifications
| Chemical Name or Material | Vemurafenib |
| CAS | 918504-65-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H18ClF2N3O3S |
| Quantity | 10 mg |
| Synonym | PLX4032 RG7204 RO5185426 |
| Solubility Information | DMSO : 50 mg/mL (102.06 mM; Need ultrasonic) |
| SMILES | FC1=CC=C(C(F)=C1C(C2=CNC3=NC=C(C=C32)C4=CC=C(C=C4)Cl)=O)NS(CCC)(=O)=O |
| Molecular Weight (g/mol) | 489.92 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction