missing translation for 'onlineSavingsMsg'
Learn More

Veledimex, MedChemExpress

Product Code. 16433217
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16433217 50 mg 50mg
16419727 5 mg 5mg
16417198 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16433217 Supplier Medchem Express Supplier No. HY16785

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Veledimex (INXN-1001), a synthetic analog of the insect molting hormone ecdysone, is an orally active activator ligand for a proprietary gene therapy promoter system. Veledimex can be used to activate certain genes using the ecdysone receptor (EcR)-based inducible gene regulation system, the RheoSwitch Therapeutic System (RTS). Veledimex can cross blood-brain barrier (BBB) in both orthotopic GL-261 mice and cynomolgus monkeys.

Veledimex (INXN-1001), a synthetic analog of the insect molting hormone ecdysone, is an orally active activator ligand for a proprietary gene therapy promoter system. Veledimex can be used to activate certain genes using the ecdysone receptor (EcR)-based inducible gene regulation system, the RheoSwitch Therapeutic System (RTS). Veledimex can cross blood-brain barrier (BBB) in both orthotopic GL-261 mice and cynomolgus monkeys.

Specifications

Chemical Name or Material Veledimex
CAS 1093130-72-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H38N2O3
Quantity 50 mg
Synonym INXN-1001 RG-115932
Solubility Information DMSO : 25 mg/mL (57.00 mM; Need ultrasonic)
SMILES O=C(NN(C(C1=CC(C)=CC(C)=C1)=O)[C@@H](C(C)(C)C)CCC)C2=CC=CC(OC)=C2CC
Molecular Weight (g/mol) 438.6
Formula Weight 438.6
Percent Purity 98.45%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.