missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFR3/Flt-4 Antibody (AFL4), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43259H
This item is not returnable.
View return policy
Description
VEGFR3/Flt-4 Monoclonal antibody specifically detects VEGFR3/Flt-4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Dual RNAscope ISH-IHC
Specifications
| VEGFR3/Flt-4 | |
| Monoclonal | |
| HRP | |
| EC 2.7.10, EC 2.7.10.1, FLT-4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4 | |
| The immunogen for this antibody was VEGFR3. | |
| 0.1 mL | |
| Angiogenesis, Cancer, Hypoxia, Protein Kinase | |
| 2324 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot | |
| AFL4 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction