missing translation for 'onlineSavingsMsg'
Learn More

VEGFR3/Flt-4 Antibody (7A3), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18430424 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430424 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430424 Supplier Novus Biologicals Supplier No. NBP118653AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

VEGFR3/Flt-4 Monoclonal antibody specifically detects VEGFR3/Flt-4 in Mouse samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGFR3/Flt-4
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 7A3
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.10, EC 2.7.10.1, FLT-4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4
Host Species Rat
Immunogen Purified, insect cell-derived recombinant mouse soluble FLT-4 (120 kDa)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Hypoxia, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2324
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.