missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFR2/KDR/Flk-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£141.00
Specifications
| Antigen | VEGFR2/KDR/Flk-1 |
|---|---|
| Concentration | 0.1 mg/mL |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
VEGFR2/KDR/Flk-1 Polyclonal specifically detects VEGFR2/KDR/Flk-1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| VEGFR2/KDR/Flk-1 | |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Human, Rat, Mouse | |
| CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, FLK-1, FLK1tyrosine kinase growth factor receptor, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGFR, VEGFR2, VEGFR-2 | |
| KDR | |
| Primary | |
| Store at 4C. Do not freeze. |
| 0.1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Stem Cell Markers, Tyrosine Kinases | |
| P35918 | |
| 3791 | |
| IgG | |
| Affinity Purified | |
| This antibody reacts with a 180 kDa protein, known as Flk-1, KDR or vascular endothelial growth factor receptor 2 (VEGFR2). Flk-1 is a cell membrane receptor kinase. Flk-1 is a high affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title