missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFR1/R2/R3 Antibody (FLT1/1663) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
VEGFR1/R2/R3 Monoclonal antibody specifically detects VEGFR1/R2/R3 in Human samples. It is validated for Flow Cytometry, Immunofluorescence
Specifications
Specifications
| Antigen | VEGFR1/R2/R3 |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | FLT1/1663 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml |
| Formulation | 10mM PBS |
| Host Species | Mouse |
| Immunogen | Recombinant human VEGFR1/R2/R3 protein (Uniprot: P17948) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?