missing translation for 'onlineSavingsMsg'
Learn More

VEGFR1/R2/R3 Antibody (FLT1/1663), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18364486 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364486 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364486 Supplier Novus Biologicals Supplier No. NBP314285AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

VEGFR1/R2/R3 Monoclonal antibody specifically detects VEGFR1/R2/R3 in Human samples. It is validated for Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGFR1/R2/R3
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone FLT1/1663
Conjugate Alexa Fluor 488
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Recombinant human VEGFR1/R2/R3 protein (Uniprot: P17948)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2321
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.