missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFR1/Flt-1 Antibody (EWF), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1006G
This item is not returnable.
View return policy
Description
VEGFR1/Flt-1 Monoclonal specifically detects VEGFR1/Flt-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| VEGFR1/Flt-1 | |
| Monoclonal | |
| DyLight 488 | |
| P17948 | |
| FLT1 | |
| Recombinant human soluble Flt-1 (D5) | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, HIF Target Genes, Hypoxia, Phospho Specific, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases | |
| 2321 | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| EWF | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This will recognize the recombinant and naturally occurring form of human VEGFR1/Flt1. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction