missing translation for 'onlineSavingsMsg'
Learn More

VEGFR1/Flt-1 Antibody, CoraFluor™ 1, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-527CL1

Product Code. 30141108

  • £780.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

VEGFR1/Flt-1 Polyclonal antibody specifically detects VEGFR1/Flt-1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VEGFR1/Flt-1
Polyclonal
PBS
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CoraFluor 1
EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1
A synthetic peptide made to an internal region of the mouse VEGF Receptor 1 protein (between residues 800-900) [Uniprot: P35969]. There is 87% identity between the immunogen used for this production and the VEGF Receptor 2 protein.
0.1 mL
Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, HIF Target Genes, Hypoxia, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases
2321
Store at 4°C in the dark. Do not freeze.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.