missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VEGFD Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148886
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family and is active in angiogenesis, lymphangiogenesis, and endothelial cell growth. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-2 and VEGFR-3 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor C.
Specifications
| VEGFD | |
| Polyclonal | |
| Unconjugated | |
| VEGFD | |
| AI325264; c-fos induced growth factor; c-fos induced growth factor (vascular endothelial growth factor D); c-fos-induced growth factor; FIGF; Vascular endothelial growth factor D; vascular endothelial growth factor-D; VEGF4; VEGFD; VEGF-D; VEGFR2 and -3 ligand | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 14205 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P97946 | |
| VEGFD | |
| E. coli-derived recombinant mouse VEGF-D, Phe98-Ser206. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction