missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VEGFC Monoclonal Antibody (D3), Invitrogen™

Mouse Monoclonal Antibody
£161.00 - £322.00
Specifications
| Antigen | VEGFC |
|---|---|
| Clone | D3 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17989732
![]() |
Invitrogen™
MA550861 |
200 μL |
£322.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17959022
|
Invitrogen™
MA550862 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family, is active in angiogenesis and endothelial cell growth, and can also affect the permeability of blood vessels. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-3 receptors. Only the fully processed form can bind and activate VEGFR-2 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor D.Specifications
| VEGFC | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| AW228853; Flt4 ligand; FLT4 ligand DHM; Flt4-L; LMPH1D; Vascular endothelial growth factor C; vascular endothelial growth factor C isoform 129; vascular endothelial growth factor C isoform 184; vascular endothelial growth factor-related protein; VEGF3; VEGFC; VEGF-C; VRP | |
| VEGFC | |
| IgG2b κ | |
| Protein A/G | |
| VEGFC |
| D3 | |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| O35757, P49767 | |
| 114111, 7424 | |
| Recombinant Rat VEGFC protein, Ala108-Arg223 (Accession #O35757). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title